Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium trifluoromethanesulfonate, 99% min

Catalog No. AA4013418
Click to view available options
Quantity:
10 g
50 g
100 g

CAS: 55120-75-7 | C2CaF6O6S2 | 338.204 g/mol

Calcium trifluoromethanesulfonate is used for aminolysis of epoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55120-75-7
Molecular Formula C2CaF6O6S2
Molecular Weight (g/mol) 338.204
MDL Number MFCD00143911
InChI Key PUQLFUHLKNBKQQ-UHFFFAOYSA-L
Synonym calcium trifluoromethanesulfonate, calcium triflate, methanesulfonic acid, trifluoro-, calcium salt, calcium trifluoromethansulphonate, methanesulfonic acid, 1,1,1-trifluoro-, calcium salt 2:1, calcium ditriflate, calcium trifluoromethyl sulfonate, calcium ii triflate, calcium 2+ ditriflate
PubChem CID 2734307
IUPAC Name calcium;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Ca+2]

Specifications

Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 50 g
Linear Formula Ca(OSO2CF3)2
Solubility Information Soluble in water.
Formula Weight 338.21
Percent Purity ≤99%
Chemical Name or Material Calcium trifluoromethanesulfonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 232-188-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.