Alfa Aesar™ Calcium tungsten oxide, 98%

Manufacturer: Alfa Aesar™ 1305136

Quantity 500g

 View more versions of this product

Catalog No. AA1305136

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7790-75-2
    Formula Weight 287.93
    MDL Number MFCD00010913
    Merck Index 14,1712
    Synonym Calcium tungstate
    Chemical Name or Material Calcium tungsten oxide

    Specifications

    Formula Weight 287.93
    Solubility Information Insoluble in H2O; Decomposes by hot HCl, HNO3
    Mesh Size −325 Mesh
    MDL Number MFCD00010913
    Melting Point 1620°C
    TSCA TSCA
    Physical Form Powder
    CAS 7790-75-2
    Merck Index 14,1712
    Quantity 500g
    Synonym Calcium tungstate
    Chemical Name or Material Calcium tungsten oxide
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit