Learn More
Invitrogen™ CALCOCO1 Polyclonal Antibody

Description
Peptide sequence: ESTTDGSPIH TSVQFQASYL PKPGAQLYQF RYVNRQGQVC GQSPPFQFRE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Specifications
Specifications
| Antigen | CALCOCO1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | Calcoco1 |
| Gene Accession No. | Q9P1Z2 |
| Gene Alias | 1810009B06Rik; calcium binding and coiled coil domain 1; calcium binding and coiled-coil domain 1; calcium-binding and coiled-coil domain-containing protein 1; Calcoco1; Calphoglin; Cocoa; Coiled-coil coactivator protein; coiled-coil leucine zipper coactivator 1; coiled-coil transcriptional coactivator; Gcap11; granule cell antiserum positive 11; GRIP1-interacting protein; inorganic pyrophosphatase activator; KIAA1536; MGC128949 protein; mKIAA1536; PP13275; Sarcoma antigen NY-SAR-3; UNQ2436/PRO4996 |
| Gene Symbols | Calcoco1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.