Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CALCOCO2 Monoclonal Antibody (OTI2D11), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501972S


Description
CALCOCO2 Monoclonal antibody specifically detects CALCOCO2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
CALCOCO2 | |
Monoclonal | |
0.53 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q13137 | |
Mouse | |
Affinity chromatography | |
RUO | |
10241 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI2D11 | |
Unconjugated | |
CALCOCO2 | |
2410154J16Rik; antigen nuclear dot 52 kDa protein; C77254; calcium binding and coiled-coil domain 2; calcium-binding and coiled-coil domain-containing protein 2; CALCOCO2; Ndp52; Ndp52l1; nuclear domain 10 protein 52; nuclear domain 10 protein 52 like 1; nuclear domain 10 protein NDP52; Nuclear dot protein 52; RGD1559974 | |
Full length human recombinant protein of human CALCOCO2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction