Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CALCOCO2 Monoclonal Antibody (P1813)

Catalog No. PIMA518293
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518293 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518293 Supplier Invitrogen™ Supplier No. MA518293
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: Huh-7, Hep3B, HepG2, Jurkat, Raji, NCI-H929, Huh-7 (untreated), Huh-7 (3 μM Thapsigargin treatment for 12 hr), Huh-7 (3 μM Thapsigargin treatment for 24 hr), A.HepG2 (untreated), HepG2 (3 μM Thapsigargin treatment for 12 hr), HepG2 (3 μM Thapsigargin treatment for 24 hr). Predicted reactivity: Human (99%), Chimpanzee (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a subunit of nuclear domain 10 (ND10) bodies. ND10 bodies are nuclear domains appearing immunohistochemically as ten dots per nucleus. They are believed to be associated with the nuclear matrix on the basis of their resistance to nuclease digestion and salt extraction. ND10 proteins are removed from the nucleus by herpes simplex virus-1 infection and may have a role in viral life cycles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CALCOCO2
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone P1813
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CALCOCO2
Gene Accession No. A2A6M5, Q13137
Gene Alias 2410154J16Rik; antigen nuclear dot 52 kDa protein; C77254; calcium binding and coiled-coil domain 2; calcium-binding and coiled-coil domain-containing protein 2; CALCOCO2; Ndp52; Ndp52l1; nuclear domain 10 protein 52; nuclear domain 10 protein 52 like 1; nuclear domain 10 protein NDP52; Nuclear dot protein 52; RGD1559974
Gene Symbols CALCOCO2
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human NDP52. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10241, 76815
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.