Learn More
CALCOCO2 Mouse anti-Canine, Human, Rat, Clone: OTI2D11, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501972

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a subunit of nuclear domain 10 (ND10) bodies. ND10 bodies are nuclear domains appearing immunohistochemically as ten dots per nucleus. They are believed to be associated with the nuclear matrix on the basis of their resistance to nuclease digestion and salt extraction. ND10 proteins are removed from the nucleus by herpes simplex virus-1 infection and may have a role in viral life cycles.Specifications
CALCOCO2 | |
Monoclonal | |
Unconjugated | |
CALCOCO2 | |
NDP52 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10241, 303479, 480545 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI2D11 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q13137 | |
CALCOCO2 | |
Full length human recombit protein of human CALCOCO2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.