Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CALCOCO2 Mouse anti-Human, Mouse, Rat, Clone: 2F2C2, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 664011IG150UL

Description
The protein encoded by this gene is a subunit of nuclear domain 10 (ND10) bodies. ND10 bodies are nuclear domains appearing immunohistochemically as ten dots per nucleus. They are believed to be associated with the nuclear matrix on the basis of their resistance to nuclease digestion and salt extraction. ND10 proteins are removed from the nucleus by herpes simplex virus-1 infection and may have a role in viral life cycles.Specifications
CALCOCO2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2A6M5, Q13137 | |
Calcoco2 | |
CALCOCO2 Fusion Protein Ag2866 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
2F2C2 | |
Unconjugated | |
CALCOCO2 | |
CALCOCO2, NDP52, Nuclear domain 10 protein 52, Nuclear dot protein 52 | |
Mouse | |
Protein A | |
RUO | |
10241, 303479, 76815 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction