Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CALCOCO2 Recombinant Rabbit Monoclonal Antibody (10Q0T10)
GREENER_CHOICE

Catalog No. FEMA551479
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551479 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551479 Supplier Invitrogen™ Supplier No. MA551479
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MEETIKDPPT SAVLLDHCHF SQVIFNSVEK FYIPGGDVTC HYTFTQHFIP RRKDWIGIFR VGWKTTREYY TFMWVTLPID LNNKSAKQQE VQFKAYYLPK DDEYYQFCYV DEDGVVRGAS IPFQFRPENE EDILVVTTQG EVEEIEQHNK ELCKENQELK DSCISLQKQN SDMQAELQKK QEELETLQSI NKKLELKVKE QKDYWETELL QLKEQNQKMS SENEKMGIRV DQLQAQLSTQ EKEMEKLVQG DQDKTEQLEQ LKKENDHLFL SLTEQRKDQK KLE.

The protein encoded by this gene is a subunit of nuclear domain 10 (ND10) bodies. ND10 bodies are nuclear domains appearing immunohistochemically as ten dots per nucleus. They are believed to be associated with the nuclear matrix on the basis of their resistance to nuclease digestion and salt extraction. ND10 proteins are removed from the nucleus by herpes simplex virus-1 infection and may have a role in viral life cycles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CALCOCO2
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 10Q0T10
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene CALCOCO2
Gene Accession No. Q13137
Gene Alias 2410154J16Rik; antigen nuclear dot 52 kDa protein; C77254; calcium binding and coiled-coil domain 2; calcium-binding and coiled-coil domain-containing protein 2; CALCOCO2; Ndp52; Ndp52l1; nuclear domain 10 protein 52; nuclear domain 10 protein 52 like 1; nuclear domain 10 protein NDP52; Nuclear dot protein 52; RGD1559974
Gene Symbols CALCOCO2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-283 of human NDP52 (NP_005822.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10241
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.