Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CALCRL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533382
Description
CALCRL Polyclonal Antibody for IHC (P)
CRLR is a receptor for calcitonin gene-related peptide type 1. The activity of CRLR is mediated by G proteins which activate adenylyl cyclase. CRLR expression has been reported in brain, lung, blood vessel, liver, and intestinal tract. ESTs have been isolated from B-cell/lung/testis, bome marrow, embryo, lung and synovium libraries.
Specifications
| CALCRL | |
| Polyclonal | |
| Unconjugated | |
| CALCRL | |
| AV071593; Calcitonin gene-related peptide type 1 receptor; calcitonin receptor like receptor; calcitonin receptor-like; calcitonin receptor-like receptor; CALCRL; CGRP receptor; CGRP type 1 receptor; Cgrp1 receptor; CGRPR; CLR; CRLR; RATCRLR; RNCLR | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100054281, 100354426, 100753229, 100861156, 10203, 25029, 397172, 488438, 527140 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| A6QP74, Q16602, Q63118, Q8WN93 | |
| CALCRL | |
| Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human CALCRL. | |
| 50 μg | |
| Primary | |
| Human, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Frog, Goat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction