Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calcyclin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517001
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517001 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517001 Supplier Invitrogen™ Supplier No. PA517001
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

Calcyclin is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in stimulation of Ca2+-dependent insulin release, stimulation of prolactin secretion, and exocytosis. Chromosomal rearrangements and altered expression of this gene have been implicated in melanoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calcyclin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 48.2 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene Cacybp
Gene Accession No. Q6AYK6, Q9CXW3, Q9HB71
Gene Alias 2A9; 5B10; CABP; CACY; CacyBP; Calcyclin; calcyclin binding protein; calcyclin-binding protein; GIG5; growth-inhibiting gene 5 protein; hCacyBP; PNAS-107; PRA; RP1-102G20.6; S100A6; S100A6-binding protein; S100A6BP; Siah-interacting protein; Siah-interacting protein (SIP); SIP
Gene Symbols Cacybp
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding Trp162 of human CACYBP.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12301, 27101, 289144
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.