Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAPS2 Monoclonal Antibody (3C6)
GREENER_CHOICE

Catalog No. PIMA524499
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524499 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524499 Supplier Invitrogen™ Supplier No. MA524499
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: YRKSYVRKAF MKLDFNKSGS VPIINIRKCY CAKKHSQVIS GHSTEEEIKS SFLETLKVAC SKSDEVSYGE FEDYYEGLSI GIVDDEDFVN ILRTPWGI.

Calcyphosine-2 is a calcium-binding protein with 2 EF-hand motifs (Wang et al., 2002 [PubMed 11846421]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAPS2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3C6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene CAPS2
Gene Accession No. Q9BXY5
Gene Alias calcyphosin 2; calcyphosin-2; calcyphosine 2; calcyphosine-2; calcyphosphine 2; CAPS2; D630005B03Rik; UG0636c06
Gene Symbols CAPS2
Host Species Mouse
Immunogen CAPS2 (NP_115995.1, 285 a.a. ~ 382 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84698
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.