Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CAPS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551218
Description
The antibody detects endogenous levels of total CAPS2 protein.
Calcyphosine-2 is a calcium-binding protein with 2 EF-hand motifs (Wang et al., 2002 [PubMed 11846421]).
Specifications
CAPS2 | |
Polyclonal | |
Unconjugated | |
CAPS2 | |
calcyphosin 2; calcyphosin-2; calcyphosine 2; calcyphosine-2; calcyphosphine 2; CAPS2; D630005B03Rik; UG0636c06 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84698 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9BXY5 | |
CAPS2 | |
Fusion protein of human CAPS2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction