Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caldesmon/CALD1 Antibody (CALD1/820 + h-CALD) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2478190.2MG
Description
Caldesmon/CALD1 Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
Caldesmon/CALD1 | |
Monoclonal | |
1.0 mg/mL | |
Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.25 - 0.5 ug/ml, Immunofluorescence : 1 - 2 ug/ml, CyTOF-ready | |
CAD, caldesmon, caldesmon 1, CDMH-CAD, HCAD, LCAD, L-CAD, MGC21352, NAG22 | |
Mouse | |
150 kDa | |
0.2 mg | |
Cytoskeleton Markers | |
800 | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF | |
CALD1/820 + h-CALD | |
Unconjugated | |
PBS with No Preservative | |
CALD1 | |
Recombinant full-length human CALD1 protein (CALD1/820); Crude human uterus extract (h-CALD) | |
Protein A or G purified | |
RUO | |
Primary | |
Recognizes a protein of 150kDa, which is identified as the high molecular weight variant of Caldesmon. Two closely related variants of human caldesmon have been identified which are different in their electrophoretic mobility and cellular distribution. The h-caldesmon variant (120-150kDa) is predominantly expressed in smooth muscle whereas l-caldesmon (70-80kDa) is found in non- muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle. This MAb recognizes only the 150kDa variant (h-caldesmon) in Western blots of human aortic media extracts and is unreactive with fibroblast extracts from cultivated human foreskin. Caldesmon is a developmentally regulated protein involved in smooth muscle and non-muscle contraction. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction