Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Caldesmon/CALD1 Antibody (SPM168), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247817MFV450
Description
Caldesmon/CALD1 Monoclonal antibody specifically detects Caldesmon/CALD1 in Human samples. It is validated for Flow CytometrySpecifications
Caldesmon/CALD1 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
SPM168 | |
Flow Cytometry | |
CAD, caldesmon, caldesmon 1, CDMH-CAD, HCAD, LCAD, L-CAD, MGC21352, NAG22 | |
Crude human uterus extract | |
0.1 mL | |
Cell Biology, Cellular Markers, Phospho Specific | |
800 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction