Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caldesmon Monoclonal Antibody (l-CALD), Invitrogen™

Catalog No. PIMA511777
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA511777 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA511777 Supplier Thermo Scientific Supplier No. MA511777
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-11777 targets Caldesmon LMW in IF, IHC (P), and WB applications and shows reactivity with Human samples. The MA5-11777 immunogen is recombinant caldesmon.

Caldesmon is a developmentally regulated protein involved in smooth muscle and non muscle contraction. Two closely related variants of human caldesmon have been identified which differ in their electrophorectic mobility and cellular distribution. The h caldesmon variant (120 - 150 kDa) is predominantly expressed in smooth muscle whereas l caldesmon (70 - 80 kDa) is found in non-muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Caldesmon
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone l-CALD
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene Cald1
Gene Accession No. Q05682
Gene Alias 4833423D12Rik; AI195384; AV071549; AW536160; C920027I18Rik; CAD; Cald1; Caldesmon; caldesmon 1; CDM; HCAD; H-CAD; h-caldesmon; LCAD; L-CAD; L-caldesmon; MGC21352; NAG22; Non-muscle caldesmon; OTTHUMP00000207950; OTTHUMP00000207953; OTTHUMP00000207954; OTTHUMP00000207955; OTTHUMP00000214473; smooth muscle caldesmon; testis secretory sperm-binding protein Li 227n
Gene Symbols Cald1
Host Species Mouse
Immunogen Recombinant caldesmon
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109624, 800
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.