Learn More
Honeywell Analytics™ CalGas Calibration Gases
Choose from both multigas mixtures and single-gas calibration gases
Supplier: Honeywell Analytics™ 549034
Description
- Calibrates gas detectors to ensure accuracy
- Multigas mixtures presented are not for use with CO+ or Tox1 sensors
Order Info
- Among single-gas calibration gases, Cat. Nos. 19-806-863 and 19-806-880 are for Mighty-Tox series bump test only; 19-806-865 is for use with Biosystems 12-022 regulator (19-806-687) only.
- For information about additional gases not offered here, contact your Fisher Safety Customer Service Representative at 1-800-772-6733.
Specifications
15.32 gal. | |
25ppm | |
Calibration Gas |
58 L | |
Hydrogen Sulfide | |
For mighty-tox series bump test only |
Safety and Handling
ShelfLife : 1 Year