Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Calhex 231 hydrochloride
GREENER_CHOICE

Catalog No. 438710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-871-0 10 mg
43-875-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-871-0 Supplier Tocris Bioscience™ Supplier No. 4387/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Negative allosteric modulator of calcium-sensing receptor (CaSR)

Negative allosteric modulator of the calcium sensing receptor (CaSR). Blocks increases in [3H]inositol phosphate levels elicited by wild-type hCasR activation (IC50= 0.39 μM in transiently transfected HEK293 cells).

Chemical Identifiers

CAS 652973-93-8
Molecular Formula C25H27ClN2O

Specifications

Quantity 10 mg
Formula Weight 443.41
Purity >98%
Molecular Weight (g/mol) 406.954
Chemical Name or Material Calhex 231 hydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.