Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CALHM1 Mouse anti-Human,Mouse, Unconjugated, Clone: 32C2, MilliporeSigma™

Catalog No. MABN25211MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN25211MI 100 μL
MABN25212MI 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN25211MI Supplier MilliporeSigma Supplier No. MABN2521100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CALHM1 Monoclonal antibody specifically detects CALHM1 in Human,Mouse samples. It is validated for Dot Blot, Immunohistochemistry, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CALHM1
Applications Dot Blot, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 32C2
Conjugate Unconjugated
Gene Accession No. NP_001001412.3
Gene Alias Calcium homeostasis modulator protein 1;Protein FAM26C
Gene Symbols CALHM1, FAM26C
Host Species Mouse
Immunogen Recombinant fragment corresponding to 144 amino acids from the C-terminal region of human Calcium homeostasis modulator protein 1 (CALHM1).
Molecular Weight of Antigen ∼46 kDa observed; 38.26 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 100 μL
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone 32C2 is a mouse monoclonal antibody that detects Calcium homeostasis modulator protein 1 (CALHM1). It targets an epitope within 11 amino acids from the C-terminal region.
Target Species Human, Mouse
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.