Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CALHM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553779
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553779 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553779 Supplier Invitrogen™ Supplier No. PA553779
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CTEHAKAFAK VCIQQFFEAM NHDLELGHTH GTLATAPASA AAPTTPDGAE EEREKLRGIT DQGTMNRLLT SWHKCKPPLR LGQEEPPLMG NGW Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 80%.

Alzheimers disease (AD) is a progressive neurodegenerative disorder characterized by a massive loss of neurons in several brain regions and by the presence of senile plaques comprised of aggregated amyloid-beta (Abeta) peptides produced by the endoproteolysis of the amyloid precursor protein (APP). The calcium homeostasis modulator 1 (CALHM1) is a recently identified multipass transmembrane glycoprotein that controls cytosolic Ca2+ concentration and Abeta levels. While some studies suggest CALHM1 is significantly associated with risk of AD, other reports suggest there is no link. CALHM1 has been suggested to modulate the Ca2+-dependent proteolytic process of APP through controlling the level of cytosolic Ca2+.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CALHM1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Calhm1
Gene Accession No. Q8IU99
Gene Alias calcium homeostasis modulator 1; calcium homeostasis modulator protein 1; Calhm1; EG546729; Fam26c; family with sequence similarity 26, member C; Protein FAM26C; RGD1564891
Gene Symbols Calhm1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CALHM1. Recombinant protein control fragment (Product #RP-91132).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255022
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.