Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CALHM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553219
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553219 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553219 Supplier Invitrogen™ Supplier No. PA553219
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RGEAYVCALS EFVDPSSLTA REEHFPSAHA TEILARFPCK ENPDNLSDFR EEVSRRLRYE SQ Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 84%.

CALHM2, also named as FAM26B, is a pore-forming subunit of a voltage-gated ion channel. It is a critical ATP-releasing channel that modulates neural activity and as a potential risk factor of depression. There are three isoforms of CALHM2 with calculated MW 36 kDa, 24 kDa and 22 kDa. It is about 32-36 kDa in western blot with membrane protein extracted lysate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CALHM2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Calhm2
Gene Accession No. Q5RJQ8, Q8VEC4, Q9HA72
Gene Alias 2810048G17Rik; calcium homeostasis modulator 2; calcium homeostasis modulator protein 2; CALHM2; Fam26b; family with sequence similarity 26, member B; Protein FAM26B; RGD1308276
Gene Symbols Calhm2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CALHM2. Recombinant protein control fragment (Product #RP-91152).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 294019, 51063, 72691
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.