Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics Calibrators for cobas c 111 Chemistry Analyzer

Catalog No. 22045812 Shop All Roche Diagnostics Products
Change view
Click to view available options
Type:
CFAS Calibrator
ISE Calibrator Kit
Quantity:
6 x 8 mL
12 x 3 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
22-045-812 CFAS Calibrator 12 x 3 mL
22-045-803 ISE Calibrator Kit 6 x 8 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-045-812 Supplier Roche Diagnostics Supplier No. 10759350360

Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the Fisher Scientific channel.

View Alternate Product

Specifications

For Use With (Equipment) Roche Cobas™ c 111 chemistry analyzer
Type CFAS Calibrator
Includes Tetramethylammonium Chloride
pH 6.9
Quantity 12 x 3 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.