Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roche Diagnostics Calibrators for cobas c 111 Chemistry Analyzer

Catalog No. p-3592455 Shop All Roche Diagnostics Products
Click to view available options
Type:
CFAS Calibrator
ISE Calibrator Kit
Quantity:
6 x 8 mL
12 x 3 mL

Specifications

For Use With (Equipment) Roche Cobas™ c 111 chemistry analyzer
Type CFAS Calibrator
Includes Tetramethylammonium Chloride
pH 6.9
Quantity 12 x 3 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.