Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calicivirus Antibody (FCV1-43) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$214.50 - $518.50
Specifications
Antigen | Calicivirus |
---|---|
Clone | FCV1-43 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Immunohistochemistry, Western Blot, ELISA, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017548
![]() |
Novus Biologicals
NB100632860.025MG |
0.025 mg |
Each for $214.50
|
|
|||||
NB10063286
![]() |
Novus Biologicals
NB10063286 |
0.125 mg |
Each for $518.50
|
|
|||||
Description
Calicivirus Monoclonal specifically detects Calicivirus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Calicivirus | |
1.0 mg/mL | |
Immunohistochemistry, Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Virus | |
Calcivirus viral capsid protein (63 kDa) | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
FCV1-43 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
FCV | |
IgG2a | |
Protein A purified | |
NB100-63286 specifically recognizes feline calicivirus, and does not cross-react with FIV or FeLV.The antibody detects a 63kD band in Western blots, representing the viral capsid protein. It may also be used in immunofluorescence techniques to detect viral infection, and has neutralising activity against feline calicivirus. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title