Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calicivirus Antibody (FCV1-43), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10063286V
Description
Calicivirus Monoclonal specifically detects Calicivirus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Calicivirus | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
NB100-63286 specifically recognizes feline calicivirus, and does not cross-react with FIV or FeLV.The antibody detects a 63kD band in Western blots, representing the viral capsid protein. It may also be used in immunofluorescence techniques to detect viral infection, and has neutralising activity against feline calicivirus. | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FCV1-43 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
FCV | |
Calcivirus viral capsid protein (63 kDa) | |
0.125 mL | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction