Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CALM Polyclonal Antibody
CALM Polyclonal Antibody
Supplier: Thermo Scientific PA531344
Description
CALM Polyclonal Antibody for Western Blot, IHC (P)

Specifications
CALM | |
Polyclonal | |
Unconjugated | |
Picalm | |
CALM; Clathrin assembly lymphoid myeloid leukemia; clathrin assembly lymphoid myeloid leukemia protein; clathrin-assembly lymphoid leukemia protein; clathrin-assembly lymphoid myeloid leukemia protein; CLTH; fit1; fit-1; LAP; mKIAA4114; PAP180; phosphatidylinositol binding clathrin assembly protein; phosphatidylinositol-binding clathrin assembly protein; PICALM; rCALM | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8301 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q13492 | |
Picalm | |
Recombinant fragment corresponding to a region within amino acids 1 and 307 of Human CALM. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction