Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CALML3 Antibody (004), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290114APC
Description
CALML3 Monoclonal antibody specifically detects CALML3 in Human samples. It is validated for ELISASpecifications
CALML3 | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
004 | |
ELISA | |
calmodulin-like 3, calmodulin-like protein 3, CaM-like protein, CLPCalmodulin-related protein NB-1 | |
Rabbit immunized with purified, recombinant Human CALML3 (Uniprot#: P27482, Met1-Lys149) | |
0.1 mL | |
Signal Transduction | |
810 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction