Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CALML3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560891
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560891 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560891 Supplier Invitrogen™ Supplier No. PA560891
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AELRHVMTRL GEKLSDEEVD EMIRAA Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 96%.

May function as a specific light chain of unconventional myosin-10 (MYO10), also enhances MYO10 translation, possibly by acting as a chaperone for the emerging MYO10 heavy chain protein. May compete with calmodulin by binding, with different affinities, to cellular substrates. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CALML3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CALML3
Gene Accession No. P27482
Gene Alias 2310068O22Rik; AI326174; CALML3; calmodulin like 3; calmodulin-like 3; calmodulin-like protein 3; Calmodulin-related protein NB-1; caM-like protein; CLP
Gene Symbols CALML3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CALML3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 810
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.