Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CALML3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103688
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103688 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103688 Supplier Invitrogen™ Supplier No. PA5103688
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CALML3.

May function as a specific light chain of unconventional myosin-10 (MYO10), also enhances MYO10 translation, possibly by acting as a chaperone for the emerging MYO10 heavy chain protein. May compete with calmodulin by binding, with different affinities, to cellular substrates. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CALML3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CALML3
Gene Accession No. P27482, Q5U206, Q9D6P8
Gene Alias 2310068O22Rik; AI326174; CALML3; calmodulin like 3; calmodulin-like 3; calmodulin-like protein 3; Calmodulin-related protein NB-1; caM-like protein; CLP
Gene Symbols CALML3
Host Species Rabbit
Immunogen A synthesized peptide derived from human CALML3(Accession P27482), corresponding to amino acid residues V92-F142.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 307100, 70405, 810
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.