Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CALML5 Antibody (119), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289981MFV610
Description
CALML5 Monoclonal antibody specifically detects CALML5 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
CALML5 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
119 | |
Immunohistochemistry-Paraffin | |
calmodulin-like 5, calmodulin-like protein 5, CLSPCalmodulin-like skin protein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CALML5 (Accession#: AAH39172.1; Met1-Glu146). | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication, Neurodegeneration, Neuroscience, Signal Transduction | |
51806 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction