Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CALML6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568378
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568378 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568378 Supplier Invitrogen™ Supplier No. PA568378
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CALML6 Polyclonal Antibody for IHC (P)

CALML6 (CAGLP) belongs to the calmodulin family and Calglandulin subfamily. It contains 4 EF-hand domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CALML6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CALML6
Gene Accession No. Q8TD86
Gene Alias CAGLP; Calglandulin-like protein; CALGP; CALML6; calmodulin like 6; calmodulin-like 6; calmodulin-like protein 6; EF-hand protein
Gene Symbols CALML6
Host Species Rabbit
Immunogen Synthesized peptide derived from human CALML6.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163688
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.