Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CALML6 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 222741AP150UL

Description
The antibody is specific to CALML6.
CALML6 (CAGLP) belongs to the calmodulin family and Calglandulin subfamily. It contains 4 EF-hand domains.Specifications
CALML6 | |
Polyclonal | |
Unconjugated | |
CALML6 | |
CAGLP, Calglandulin like protein, CALGP, CALML6, calmodulin like 6, Calmodulin like protein 6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
163688 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.54 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8TD86 | |
CALML6 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction