Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calmodulin 2 Antibody (J4D8), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP321504F
Description
Calmodulin 2 Monoclonal antibody specifically detects Calmodulin 2 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
Calmodulin 2 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunofluorescence | |
J4D8 | |
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
CALM, CALM1, CALM3, CALML2, calmodulin, calmodulin 2 (phosphorylase kinase, delta), CAM, CAM1, CAM2, CAM3, CAMB, CAMC, CAMII, CAMIII, FLJ99410, LP7057 protein, PHKD, PHKD2, phosphorylase kinase delta | |
Recombinant human Calmodulin (1-149aa) purified from E. coli (Accesion #: NP_001734; Uniprot #: P0DP24). | |
0.1 mL | |
Cancer | |
805 | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction