Learn More
Invitrogen™ Calmodulin 3 Monoclonal Antibody (1E2)

Description
Peptide Sequence: INEVDADGNG TIDFPEFLTM MARKMKDTDS EEEIREAFRV FDKDGNGYIS AAELRHVMTN LGEKLTDEEV DEMIREADID GDGQVNYEEF VQMMTAK.
Specifications
Specifications
| Antigen | Calmodulin 3 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1E2 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | CALM3 |
| Gene Accession No. | P0DP25 |
| Gene Alias | Calm; CALM1; Calm2; Calm3; CALML2; calmodulin; calmodulin 1; Calmodulin 1 (phosphorylase kinase, delta); calmodulin 3; calmodulin 3 (phosphorylase kinase, delta); Calmodulin III; calmodulin-1; Calmodulin-2; Calmodulin-3; Cam; Cam1; CAM2; Cam3; CaMA; CAMB; Camc; CAMI; CaMII; CAMIII; CPVT4; DD132; epididymis secretory protein Li 72; HEL-S-72; LQT14; PHKD; PHKD3; phosphorylase kinase subunit delta; phosphorylase kinase, delta subunit; prepro-calmodulin 1; prepro-calmodulin 3; R75142; RNCAMIII |
| Gene Symbols | CALM3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.