Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calmodulin 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588172
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588172 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588172 Supplier Invitrogen™ Supplier No. PA588172
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse brain; Cellular Location: Cytoplasm, cytoskeleton, spindle, spindle pole Immunogen sequence: MADQLTEEQI AEFKEAFSLF DKDGDGTITT KELGTVMRSL GQNPTEAELQ DMINEVDADG NGTIDFPEFL TMMARKMKDT DSEEEIREAF RVFDKDGNGY ISAAELRHVM TNLGEKLTDE EVDEMIREAD IDGDGQVNYE EFVQMMTAK.

Calcium binding proteins (CaBP) play a crucial role in the calcium-mediated cellular signal transduction pathway in the central nervous system. The CaBP family shares much similarity with CaM I (calmodulin), and it has been shown that CaBP proteins can substitute functionally for, and possibly augment the function of, CaM I. CaBP7 (Calcium-binding protein 7), contains two EF-hand domains for calcium binding and shares 70% homology with CaBP8 and 50% homology with CaM I. It negatively regulates Golgi-to-plasma membrane trafficking by interacting with PI4KB and inhibiting its activity. CaBP7 and 8 possess a targeting mechanism that is unique amongst the CaBPs that may contribute to differential functional Ca2+-sensing by these family members.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calmodulin 3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 5.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CALM3
Gene Accession No. P0DP25, P0DP28
Gene Alias Calm; CALM1; Calm2; Calm3; CALML2; calmodulin; calmodulin 1; Calmodulin 1 (phosphorylase kinase, delta); calmodulin 3; calmodulin 3 (phosphorylase kinase, delta); Calmodulin III; calmodulin-1; Calmodulin-2; Calmodulin-3; Cam; Cam1; CAM2; Cam3; CaMA; CAMB; Camc; CAMI; CaMII; CAMIII; CPVT4; DD132; epididymis secretory protein Li 72; HEL-S-72; LQT14; PHKD; PHKD3; phosphorylase kinase subunit delta; phosphorylase kinase, delta subunit; prepro-calmodulin 1; prepro-calmodulin 3; R75142; RNCAMIII
Gene Symbols CALM3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-149 of human CALM3 (NP_0017341).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12315, 808
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.