Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Calmodulin 3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589886
Description
Positive Samples: Rat testis; Cellular Location: Cytoplasm, cytoskeleton, spindle, spindle pole Immunogen sequence: LGQNPTEAEL QDMINEVDAD GNGTIDFPEF LTMMARKMKD TDSEEEIREA FRVFDKDGNG YISAAELRHV MTNLGEKLTD EEVDEMIREA DIDGDGQVNY E.
Calcium binding proteins (CaBP) play a crucial role in the calcium-mediated cellular signal transduction pathway in the central nervous system. The CaBP family shares much similarity with CaM I (calmodulin), and it has been shown that CaBP proteins can substitute functionally for, and possibly augment the function of, CaM I. CaBP7 (Calcium-binding protein 7), contains two EF-hand domains for calcium binding and shares 70% homology with CaBP8 and 50% homology with CaM I. It negatively regulates Golgi-to-plasma membrane trafficking by interacting with PI4KB and inhibiting its activity. CaBP7 and 8 possess a targeting mechanism that is unique amongst the CaBPs that may contribute to differential functional Ca2+-sensing by these family members.
Specifications
Calmodulin 3 | |
Polyclonal | |
Unconjugated | |
CALM3 | |
Calm; CALM1; Calm2; Calm3; CALML2; calmodulin; calmodulin 1; Calmodulin 1 (phosphorylase kinase, delta); calmodulin 3; calmodulin 3 (phosphorylase kinase, delta); Calmodulin III; calmodulin-1; Calmodulin-2; Calmodulin-3; Cam; Cam1; CAM2; Cam3; CaMA; CAMB; Camc; CAMI; CaMII; CAMIII; CPVT4; DD132; epididymis secretory protein Li 72; HEL-S-72; LQT14; PHKD; PHKD3; phosphorylase kinase subunit delta; phosphorylase kinase, delta subunit; prepro-calmodulin 1; prepro-calmodulin 3; R75142; RNCAMIII | |
Rabbit | |
Affinity chromatography | |
RUO | |
12315, 24244, 808 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P0DP25, P0DP28, P0DP31 | |
CALM3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 40-140 of human CALM3 (NP_0051752). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction