Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calmodulin Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. PIPA5122172
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122172 100 μL
PIPA5122173 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122172 Supplier Invitrogen™ Supplier No. PA5122172
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Calmodulin mediates the control of a large number of enzymes, ion channels, aquaporins and other proteins by Ca2+. Among the enzymes to be stimulated by the calmodulin-Ca2+ complex are a number of protein kinases and phosphatases. Together with CCP110 and centrin, is involved in a genetic pathway that regulates the centrosome cycle and progression through cytokinesis. Calmodulin is a small, highly conserved calcium binding protein found in all eukaryotic cells. With the capacity to bind up to four calcium ions, this 17 kDa protein acts as an important intracellular receptor for regulatory calcium signals. As it binds calcium, calmodulin undergoes conformational changes which can increase its affinity for target proteins. It acts both directly, through interaction with key target enzymes, and indirectly, via specific kinases. Studies have found that calmodulin participates in the regulation of several biological processes including energy and biosynthetic metabolism, cell motility, exocytosis, cytoskeletal assembly, and intracellular modulation of both cAMP and calcium concentrations. Calmodulin is the archetype of the family of calcium-modulated proteins of which nearly 20 members have been found. They are identified by their occurrence in the cytosol or on membranes facing the cytosol and by a high affinity for calcium. Calmodulin contains 149 amino acids and has 4 calcium-binding domains. Its functions include roles in growth and the cell cycle as well as in signal transduction and the synthesis and release of neurotransmitters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calmodulin
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Calm1
Gene Accession No. P0DP23, P0DP24, P0DP25
Gene Alias 1500001E21Rik; 2310037J09Rik; AI256814; AI327027; AI461935; AI649108; AL024000; AL024017; AY314008; calcium binding gene, DD112; calcium-binding protein Dd112; CALM; CALM 1; CALM1; CALM2; Calm3; Calm4; Calm5; CALML2; Calmodulin; calmodulin 1; calmodulin 1 (phosphorylase kinase, delta); calmodulin 2; calmodulin 2 (phosphorylase kinase, delta); calmodulin 3; calmodulin 3 (phosphorylase kinase, delta); calmodulin 4; calmodulin 5; Calmodulin III; calmodulin-1; Calmodulin-2; calmodulin-3; Calmodulin-4; calmodulin-like; calmodulin-like protein; calmodulin-like protein (neoCaM); caM; CAM1; Cam2; CAM3; CaMA; Camb; CamC; CaMI; CAMII; CAMIII; CPVT4; DD112; DD132; DD312; epididymis secretory protein Li 72; HEL-S-72; hypothetical protein; LP7057 protein; LQT14; LQT15; MGC128137 protein; neo-calmodulin; NeoCaM; PHKD; PHKD2; PHKD3; phosphorylase kinase delta; phosphorylase kinase subunit delta; phosphorylase kinase, delta subunit; prepro-calmodulin 1; prepro-calmodulin 2; prepro-calmodulin 3; R75142; RNCAMIII; Scarf; Scarf2; similar to calmodulin; skin calmodulin-related factor
Gene Symbols Calm1, Calm2, CALM3
Host Species Rabbit
Immunogen Met1-Lys149 (Accession P62158), with N-terminal His and GST Tag.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 801, 805, 808
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.