Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calmyrin Monoclonal Antibody (1G8C8)
GREENER_CHOICE

Catalog No. 354500
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-450-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 35-450-0 Supplier Invitrogen™ Supplier No. 354500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calmyrin Monoclonal Antibody for Western Blot, ELISA

The protein encoded by this gene is a member of the calcium-binding protein family. The specific function of this protein has not yet been determined; however this protein is known to interact with DNA-dependent protein kinase and may play a role in kinase-phosphatase regulation of DNA end joining. This protein also interacts with integrin alphabeta, which may implicate this protein as a regulatory molecule for alphabeta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calmyrin
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1G8C8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Cib1
Gene Accession No. Q99828, Q9Z0F4
Gene Alias calcium and integrin binding 1; calcium and integrin binding 1 (calmyrin); calcium and integrin binding protein 1; calcium and integrin binding, protein kinase interacting protein; Calcium- and integrin-binding protein; calcium and integrin-binding protein 1; calmyrin; CIB; CIB1; Cibkip; CIBP; DNA-dependent protein kinase interacting protein; DNA-PK interaction protein; DNA-PKcs-interacting protein; Kinase-interacting protein; KIP; KIP1; LOW QUALITY PROTEIN: calcium and integrin-binding protein 1; PRKDCIP; protein kinase, DNA activated, catalytic polypeptide interacting protein; Sip2-28; SNK-interacting protein 2-28; testicular secretory protein Li 9
Gene Symbols Cib1
Host Species Mouse
Immunogen Recombinant Calmyrin protein (full length).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10519, 23991
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.