Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Calmyrin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583931
Description
Immunogen sequence: DIKSHYAFRI FDFDDDGTLN REDLSRLVNC LTGEGEDTRL SASEMKQLID NILEESDIDR DGTINLSEFQ HVISRSPDFA SSF.
The protein encoded by this gene is a member of the calcium-binding protein family. The specific function of this protein has not yet been determined; however this protein is known to interact with DNA-dependent protein kinase and may play a role in kinase-phosphatase regulation of DNA end joining. This protein also interacts with integrin alphabeta, which may implicate this protein as a regulatory molecule for alphabeta.
Specifications
Calmyrin | |
Polyclonal | |
Unconjugated | |
Cib1 | |
calcium and integrin binding 1; calcium and integrin binding 1 (calmyrin); calcium and integrin binding protein 1; calcium and integrin binding, protein kinase interacting protein; Calcium- and integrin-binding protein; calcium and integrin-binding protein 1; calmyrin; CIB; CIB1; Cibkip; CIBP; DNA-dependent protein kinase interacting protein; DNA-PK interaction protein; DNA-PKcs-interacting protein; Kinase-interacting protein; KIP; KIP1; LOW QUALITY PROTEIN: calcium and integrin-binding protein 1; PRKDCIP; protein kinase, DNA activated, catalytic polypeptide interacting protein; Sip2-28; SNK-interacting protein 2-28; testicular secretory protein Li 9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10519 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99828 | |
Cib1 | |
Recombinant protein corresponding to Human CIB1. Recombinant protein control fragment (Product #RP-104864). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction