Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calnexin Antibody (1563), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP243765
Description
Ensure accurate, reproducible results in Western Blot
Calnexin Monoclonal specifically detects Calnexin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
Calnexin | |
Monoclonal | |
Unconjugated | |
PBS with No Preservative | |
CANX | |
Recombinant protein encompassing a sequence within the center region of human Calnexin. The exact sequence is proprietary. | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
1563 | |
Western Blot 1:500-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation | |
calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90 | |
Mouse | |
68 kDa | |
0.1 mL | |
ER Markers, Neuroscience, Phospho Specific, Sensory Systems, Unfolded Protein Response, Vision | |
821 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction