Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calnexin Antibody (CANX/1541), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254552AF350
Description
Calnexin Monoclonal specifically detects Calnexin in Human, Rat (Negative) samples. It is validated for ELISA.Specifications
Calnexin | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
CANX | |
Recombinant N-terminal fragment of human Calnexin protein (around aa 1-300) (exact sequence is proprietary) (Uniprot: P27824) | |
Primary | |
Human, Rat | |
IgG2b κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
CANX/1541 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90 | |
Mouse | |
0.1 mL | |
821 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction