Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calnexin Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1001974H
Description
Calnexin Polyclonal specifically detects Calnexin in Human, Mouse, Rat, Hamster, Zebrafish samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Calnexin | |
Polyclonal | |
Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin | |
calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90 | |
Rabbit | |
97 kDa | |
0.1 mL | |
ER Markers, Neuroscience, Sensory Systems, Unfolded Protein Response, Vision | |
821 | |
Human, Mouse, Rat, Hamster, Zebrafish | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS with No Preservative | |
CANX | |
A synthetic peptide made to a C-terminal portion of the rat Calnexin protein (between residues 550-591) [Uniprot: P35565] | |
Affinity Purified | |
RUO | |
Primary | |
Rat, human, and mouse. Hamster reactivity reported in the scientific literature (PMID: 23760268). Predicted to react with Xenopus laevis, dog, orangutan, cow, chicken, Drosophilia melanogaster, guinea pig, quail, pig, rabbit, and sheep based on 100% sequence homology. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction