Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calnexin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 11 publications
$268.00 - $706.50
Specifications
| Antigen | Calnexin |
|---|---|
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Electron Microscopy 1:10-1:500 |
| Applications | Flow Cytometry, Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018781
![]() |
Novus Biologicals
NBP1974850.025MG |
0.025 mg |
Each for $268.00
|
|
|||||
NBP19748505
![]() |
Novus Biologicals
NBP1974850.05MG |
0.05 mg |
Each for $390.00
|
|
|||||
NBP19748502
![]() |
Novus Biologicals
NBP1974850.2MG |
0.2 mg |
Each for $706.50
|
|
|||||
Description
Calnexin Polyclonal specifically detects Calnexin in Human, Mouse, Rat, Porcine, Avian, Bovine, Canine, Chicken, Drosophila, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Electron Microscopy.Specifications
| Calnexin | |
| Flow Cytometry, Western Blot, Immunofluorescence, Immunoprecipitation, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Pig, Chicken, Rat, Canine, Avian, Mouse, Drosophila, Human, Sheep, Primate, Guinea Pig, Rabbit, Hamster, Bovine | |
| P24643 | |
| 821 | |
| Synthetic peptide corresponding to the sequence near the C-terminus of canine calnexin. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Electron Microscopy 1:10-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90 | |
| CANX | |
| IgG | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title