Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calnexin Antibody, Novus Biologicals™
SDP

Goat Polyclonal Antibody has been used in 2 publications

Supplier:  Novus Biologicals NBP137774

Catalog No. NBP137774


Only null left
Add to Cart

Description

Description

Calnexin Polyclonal specifically detects Calnexin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications

Specifications

Calnexin
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90
Goat
Affinity Purified
RUO
Primary
Reported variants represent identical protein (NP_001019820.1, NP_001737.1).
Store at -20C. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ELISA
0.5 mg/mL
Western Blot 0.1 - 1 ug/mL, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 10 ug/mL, Immunohistochemistry-Paraffin 5 ug/mL, Peptide ELISA Detection limit 1:128000
P27824
CANX
Peptide with sequence C-SKTPELNLDQFHDKT corresponding to internal region (near N-Terminus) according to NP_001737.1.
0.1 mg
Vision
821
Human, Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only