Learn More
Calnexin Mouse anti-Human, Unconjugated, Clone: 2A2C6, Proteintech
Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 669031IG150UL
Description
The Calnexin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Calnexin. This antibody recognizes human, mouse, rat antigen. The Calnexin antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Calnexin | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunofluorescence, ELISA | |
2A2C6 | |
Unconjugated | |
150 μL | |
821 | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.