Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calnexin Mouse anti-Human, Unconjugated, Clone: , MilliporeSigma™

Catalog No. MABF206725 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2067-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF2067-25 Supplier MilliporeSigma Supplier No. MABF206725UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Calnexin, clone AF8, Cat. No. MABF2067, is a mouse monoclonal antibody that detects calnexin in human cells and has been tested for use in Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calnexin
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias IP90;Major histocompatibility complex class I antigen-binding protein p88;p90
Gene Symbols CANX
Host Species Mouse
Immunogen TCR complexes isolated from the human T-cell leukemia line, DND41.
Molecular Weight of Antigen 67.57 kDa calculated.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Test Specificity Clone AF8 detects Calnexin in human cells.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.