Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpain 1 Antibody (CAPN1/1530), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB386839 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB386839 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB386839 Supplier Novus Biologicals Supplier No. NBP308456IR
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calpain 1 Monoclonal antibody specifically detects Calpain 1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Calpain 1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CAPN1/1530
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias Calcium-activated neutral proteinase 1, calpain 1, (mu/I) large subunit, Calpain mu-type, calpain, large polypeptide L1, calpain-1 catalytic subunit, Calpain-1 large subunit, CANP, CANP 1, CANPL1EC 3.4.22.52, Cell proliferation-inducing gene 30 protein, cell proliferation-inducing protein 30, EC 3.4.22, Micromolar-calpain, muCANPCANP1, muCL
Host Species Mouse
Immunogen Recombinant full-length human Calpain 1 protein (Uniprot: P07384)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 823
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.