Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calpain 1 Antibody (SR2190), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32156950UL
Description
Calpain 1 Monoclonal antibody specifically detects Calpain 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
Calpain 1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
SR2190 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Calcium-activated neutral proteinase 1, calpain 1, (mu/I) large subunit, Calpain mu-type, calpain, large polypeptide L1, calpain-1 catalytic subunit, Calpain-1 large subunit, CANP, CANP 1, CANPL1EC 3.4.22.52, Cell proliferation-inducing gene 30 protein, cell proliferation-inducing protein 30, EC 3.4.22, Micromolar-calpain, muCANPCANP1, muCL | |
A synthesized peptide derived from human Calpain 1 (Uniprot #: P07384) | |
50 μg | |
Cell Cycle and Replication | |
823 | |
Store at -20 degrees C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction