Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpain 1 Monoclonal Antibody (2A1), Huabio

Catalog No. 502544819
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4819 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4819 Supplier HUABIO RESEARCH INC Supplier No. EM180102
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calpain 1 Monoclonal antibody specifically detects Calpain 1 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)

Specifications

Antigen Calpain 1
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2A1
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene CAPN1
Gene Accession No. P07384, P97571
Gene Alias calcium activated neutral protease; calcium-activated neutral protease 1; calcium-activated neutral proteinase 1; calcium-dependent protease; calpain; calpain 1; calpain 1 large subunit; calpain 1, (mu/I) large subunit; calpain 1, large subunit; calpain mu-type; calpain, large polypeptide L1; calpain-1 catalytic subunit; Calpain-1 large subunit; Calpain-I; Calpain-II; CANP; CANP 1; Canp1; CANPL1; Capa1; Capa-1; Capn1; Cell proliferation-inducing gene 30 protein; cell proliferation-inducing protein 30; Cls1; I79_024883; micromolar calcium-activated neutral protease 1; micromolar calcium-activated neutral protease 1 isoform A; micromolar calcium-activated neutral protease 1 isoform B; micromolar calcium-dependent neutral protease large subunit; micromolar-calpain; mucalpain; mu-calpain; mu-calpin; muCANP; muCL; PIG30; SPG76
Host Species Mouse
Immunogen Recombinant protein within Human Calpain 1 aa 1-714/714.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29153, 823
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.