Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calpain 1 Recombinant Rabbit Monoclonal Antibody (10E6O5)
GREENER_CHOICE

Catalog No. PIMA535705
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535705 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535705 Supplier Invitrogen™ Supplier No. MA535705
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: DGATRTDICQ GALGDCWLLA AIASLTLNDT LLHRVVPHGQ SFQNGYAGIF HFQLWQFGEW VDVVVDDLLP IKDGKLVFVH SAEGNEFWSA LLEKAYAKVN G.

The calpains including Calpain 1 (CAPN1), calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. Several transcript variants encoding two different isoforms have been found for this gene. Diseases associated with CAPN1 include Spastic Paraplegia 76, Autosomal Recessive and Spasticity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain 1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 10E6O5
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CAPN1
Gene Accession No. O35350, P07384, P97571
Gene Alias calcium activated neutral protease; calcium-activated neutral protease 1; calcium-activated neutral proteinase 1; calcium-dependent protease; calpain; calpain 1; calpain 1 large subunit; calpain 1, (mu/I) large subunit; calpain 1, large subunit; calpain mu-type; calpain, large polypeptide L1; calpain-1 catalytic subunit; Calpain-1 large subunit; Calpain-I; Calpain-II; CANP; CANP 1; Canp1; CANPL1; Capa1; Capa-1; Capn1; Cell proliferation-inducing gene 30 protein; cell proliferation-inducing protein 30; Cls1; I79_024883; micromolar calcium-activated neutral protease 1; micromolar calcium-activated neutral protease 1 isoform A; micromolar calcium-activated neutral protease 1 isoform B; micromolar calcium-dependent neutral protease large subunit; micromolar-calpain; mucalpain; mu-calpain; mu-calpin; muCANP; muCL; PIG30; SPG76
Gene Symbols CAPN1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Calpain 1 (P07384).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12333, 29153, 823
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.