Learn More
Invitrogen™ Calpain 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578911
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human Jurkat whole cell, human RT4 whole cell, human A431 whole cell, rat lung tissue, mouse lung tissue. IHC: rat lung tissue, human intestinal cancer tissue IHC-F: rat cardiac muscle tissue. ICC/IF: A549 cell, HELA Cell.
The calpains including Calpain 1 (CAPN1), calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. Several transcript variants encoding two different isoforms have been found for this gene. Diseases associated with CAPN1 include Spastic Paraplegia 76, Autosomal Recessive and Spasticity.
Specifications
Calpain 1 | |
Polyclonal | |
Unconjugated | |
CAPN1 | |
calcium activated neutral protease; calcium-activated neutral protease 1; calcium-activated neutral proteinase 1; calcium-dependent protease; calpain; calpain 1; calpain 1 large subunit; calpain 1, (mu/I) large subunit; calpain 1, large subunit; calpain mu-type; calpain, large polypeptide L1; calpain-1 catalytic subunit; Calpain-1 large subunit; Calpain-I; Calpain-II; CANP; CANP 1; Canp1; CANPL1; Capa1; Capa-1; Capn1; Cell proliferation-inducing gene 30 protein; cell proliferation-inducing protein 30; Cls1; I79_024883; micromolar calcium-activated neutral protease 1; micromolar calcium-activated neutral protease 1 isoform A; micromolar calcium-activated neutral protease 1 isoform B; micromolar calcium-dependent neutral protease large subunit; micromolar-calpain; mucalpain; mu-calpain; mu-calpin; muCANP; muCL; PIG30; SPG76 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12333, 29153, 823 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
O35350, P07384, P97571 | |
CAPN1 | |
A synthetic peptide corresponding to a sequence in the middle region of human Calpain 1 (312-326aa EWNNVDPYERDQLRV). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.